ADV412110741 AChemBlock

2-Cyano-5-(trifluoromethyl)pyridine

  • MDL number MFCD01001119
  • Cas Number : 95727-86-9
  • Molecular Weight : 172.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-cyano-5-trifluoromethyl-pyridine.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6934
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart