ADV412110749 AChemBlock

(R)-2-Methylpiperazine-1-carboxylic acid benzyl ester

  • MDL number MFCD04115051
  • Cas Number : 923565-99-5
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Physical Form yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/r-2-methylpiperazine-1-carboxylic-acid-benzyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4187
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart