ADV412110758 AChemBlock

2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD12923384
  • Cas Number : 1111638-03-9
  • Molecular Weight : 258.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-7-azaindole-5-boronic-acid-pinacol-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6028
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart