ADV412110767 AChemBlock

(1R,2R,4S)-rel-2-Amino-7-Boc-7-Azabicyclo[2.2.1]heptane

  • MDL number MFCD26961266
  • Cas Number : 1000870-15-4
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1r-2r-4s-rel-2-amino-7-boc-7-azabicyclo-2-2-1-heptane.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6961
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart