ADV412110781 AChemBlock

(2S,4R)-4-Fluoropyrrolidine-2-carboxylic acid

  • MDL number MFCD02683982
  • Cas Number : 2507-61-1
  • Molecular Weight : 133.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Physical Form pale yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2s-4r-4-fluoropyrrolidine-2-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-341
pictograms GHS07
hstatements H313;H303;H333
Availability details for
Quantity
  • Availability
  • Add to Cart