ADV412110785 AChemBlock

2-N-Fmoc-amino-3-(4-N-Boc-piperidinyl)propionic acid

  • MDL number MFCD01860501
  • Cas Number : 313052-02-7
  • Molecular Weight : 494.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-n-fmoc-amino-3-4-n-boc-piperidinyl-propionic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6979
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart