ADV412110787 AChemBlock

4-Fluoro-1H-indazole-5-carboxylic acid

  • MDL number MFCD11007930
  • Cas Number : 1041481-59-7
  • Molecular Weight : 180.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-fluoro-1h-indazole-5-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6982
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart