ADV412110794 AChemBlock

Methyl 5,5-difluoro-3-piperidinecarboxylate hydrochloride

  • MDL number MFCD20261414
  • Cas Number : 1359656-87-3
  • Molecular Weight : 215.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-5-difluoro-3-piperidinecarboxylate-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6990
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart