ADV412110799 AChemBlock

2-Chloro-5-iodo-4-pyridinol

  • MDL number MFCD19689499
  • Cas Number : 1226878-99-4
  • Molecular Weight : 255.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-5-iodo-4-pyridinol.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6995
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart