ADV412110804 AChemBlock

Benzyl 3,5-dioxopiperidine-1-carboxylate

  • MDL number MFCD16618536
  • Cas Number : 1228631-19-3
  • Molecular Weight : 247.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/benzyl-3-5-dioxopiperidine-1-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7000
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart