ADV412110819 AChemBlock

tert-Butyl 4-(3-acetylphenyl)piperidine-1-carboxylate

  • MDL number MFCD11111398
  • Cas Number : 1198283-87-2
  • Molecular Weight : 303.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-3-acetylphenyl-piperidine-1-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7017
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart