ADV412110846 AChemBlock

4-(2-bromo-4,5-dimethoxybenzyl)-4-(2-(2-(6,6-dimethylbicyclo[3.1.1]heptan-2-yl)ethoxy)ethyl)morpholin-4-ium bromide

  • MDL number MFCD01713435
  • Cas Number : 53251-94-8
  • Molecular Weight : 591.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/morpholinium-4-2-bromo-4-5-dimethoxyphenyl-methyl-4-2-2-6-6-dimethylbicyclo-3-1-1-hept-2-yl-ethoxy-ethyl-bromide-1-1.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7047
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart