ADV412110854 AChemBlock

2-Bromo-2-cyclopentylacetic acid

  • MDL number MFCD24476773
  • Cas Number : 53389-30-3
  • Molecular Weight : 207.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-2-cyclopentylacetic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4556
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart