ADV412110855 AChemBlock

Methyl 2-(difluoromethyl)-6-methoxyl-pyridine-4-carboxylate

  • MDL number MFCD18258498
  • Cas Number : 1256807-01-8
  • Molecular Weight : 217.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-difluoromethyl-6-methoxyl-pyridine-4-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7057
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart