ADV412110868 AChemBlock

tert-butyl (1R,5S)-3,6-diazabicyclo[3.2.0]heptane-6-carboxylate

  • MDL number MFCD22573659
  • Cas Number : 799279-81-5
  • Molecular Weight : 198.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-6-diazabicyclo-3-2-0-heptane-6-carboxylic-acid-1-1-dimethylethyl-ester-1r-5s.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7072
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart