ADV412110874 AChemBlock

endo-7-Hydroxy-3-oxa-9-azabicyclo[3.3.1]nonane hydrochloride

  • MDL number MFCD27956890
  • Cas Number : 240401-10-9
  • Molecular Weight : 179.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/endo-7-hydroxy-3-oxa-9-azabicyclo-3-3-1-nonane-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7082
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart