ADV412110879 AChemBlock

7-Methyl-1H-indole-5-carboxylic acid methyl ester

  • MDL number MFCD03094981
  • Cas Number : 180624-25-3
  • Molecular Weight : 189.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-methyl-1h-indole-5-carboxylic-acid-methyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7087
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart