ADV412110881 AChemBlock

3H-Spiro[2-benzofuran-1,4'-piperidine]

  • MDL number MFCD06738867
  • Cas Number : 38309-60-3
  • Molecular Weight : 189.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3h-spiro-2-benzofuran-1-4-piperidine.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4560
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart