ADV412110886 AChemBlock

2-oxoindoline-4-carbonitrile

  • MDL number MFCD20480663
  • Cas Number : 214759-51-0
  • Molecular Weight : 158.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1h-indole-4-carbonitrile-2-3-dihydro-2-oxo.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7095
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart