ADV412110888 AChemBlock

(2S,5S)-2,5-Dimethyl-piperazine dihydrobromide

  • MDL number MFCD16987704
  • Cas Number : 98778-71-3
  • Molecular Weight : 276.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2s-5s-2-5-dimethyl-piperazine-dihydrobromide.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7097
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart