ADV412110891 AChemBlock

tert-butyl (2-(aminooxy)ethyl)carbamate

  • MDL number MFCD11934722
  • Cas Number : 75051-55-7
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/carbamic-acid-2-aminooxy-ethyl-1-1-dimethylethyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4563
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart