ADV412110892 AChemBlock

7-(Trifluoromethyl)-1(2H)-isoquinolinone

  • MDL number MFCD12405863
  • Cas Number : 410086-28-1
  • Molecular Weight : 213.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-trifluoromethyl-1-2h-isoquinolinone.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7102
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart