ADV412110896 AChemBlock

N-ethyl-1-methylpiperidin-4-amine

  • MDL number MFCD07801141
  • Cas Number : 876717-32-7
  • Molecular Weight : 142.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-piperidinamine-n-ethyl-1-methyl.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7107
pictograms GHS07
hstatements H302;H313;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart