ADV412110907 AChemBlock

Diphenylvinylsulfonium triflate

  • MDL number MFCD22123284
  • Cas Number : 247129-88-0
  • Molecular Weight : 362.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/diphenylvinylsulfonium-triflate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7118
pictograms GHS07
hstatements H302;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart