ADV412110908 AChemBlock

tert-Butyl 3,5-dioxopiperidine-1-carboxylate

  • MDL number MFCD14706038
  • Cas Number : 396731-40-1
  • Molecular Weight : 213.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-5-dioxopiperidine-1-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7119
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart