ADV412110915 AChemBlock

7-methoxy-3,4-dihydroisoquinolin-1(2H)-one

  • MDL number MFCD04114863
  • Cas Number : 22246-04-4
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-2h-isoquinolinone-3-4-dihydro-7-methoxy.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7127
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart