ADV412110921 AChemBlock

3-Ethynyl-3-methyloxetane

  • MDL number MFCD19441315
  • Cas Number : 1290541-27-3
  • Molecular Weight : 96.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-ethynyl-3-methyloxetane.html
Availability details for
Quantity
  • Availability
  • Add to Cart