ADV412110930 AChemBlock

3-Boc-3-azabicyclo[3.1.0]hexane-1-carboxylic acid

  • MDL number MFCD22566159
  • Cas Number : 1363381-55-8
  • Molecular Weight : 227.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-boc-3-azabicyclo-3-1-0-hexane-1-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7143
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart