ADV412110932 AChemBlock

5-Boc-5-azaspiro[2.4]heptane-1-carboxylic acid

  • MDL number MFCD12198550
  • Cas Number : 150543-61-6
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-boc-5-azaspiro-2-4-heptane-1-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7145
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart