ADV412110937 AChemBlock

N-(Azetidin-3-yl)methanesulfonamide hydrochloride

  • MDL number MFCD20926152
  • Cas Number : 1239205-33-4
  • Molecular Weight : 186.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/n-azetidin-3-yl-methanesulfonamide-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7150
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart