ADV412110939 AChemBlock

3-Amino-5-bromo-2-ethylpyridine

  • MDL number MFCD18256168
  • Cas Number : 1093819-32-9
  • Molecular Weight : 201.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-amino-5-bromo-2-ethylpyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7153
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart