ADV412110941 AChemBlock

(S)-3-(Boc-amino)cyclopentanone

  • MDL number MFCD18089809
  • Cas Number : 167298-40-0
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-3-boc-amino-cyclopentanone.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7156
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart