ADV412110949 AChemBlock

1-(5-Bromo-2-pyrimidinyl)ethanone

  • MDL number MFCD18909512
  • Cas Number : 1189169-37-6
  • Molecular Weight : 201.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Light yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-5-bromo-2-pyrimidinyl-ethanone.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7164
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart