ADV412110954 AChemBlock

5-Bromo-2,4-dichloro-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD13193619
  • Cas Number : 900789-14-2
  • Molecular Weight : 266.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-4-dichloro-7h-pyrrolo-2-3-d-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7170
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart