ADV412110968 AChemBlock

2-Amino-7-methyl-7-azaspiro[3.5]nonane

  • MDL number MFCD12198555
  • Cas Number : 1160247-16-4
  • Molecular Weight : 154.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-amino-7-methyl-7-azaspiro-3-5-nonane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7186
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart