ADV412110971 AChemBlock

6-(Boc-amino)pyridine-3-boronic acid

  • MDL number MFCD11975411
  • Cas Number : 883231-20-7
  • Molecular Weight : 238.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-boc-amino-pyridine-3-boronic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7189
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart