ADV412110977 AChemBlock

1H-Pyrazolo[4,3-c]pyridine-7-carboxylic acid

  • MDL number MFCD16251039
  • Cas Number : 1363383-10-1
  • Molecular Weight : 163.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1h-pyrazolo-4-3-c-pyridine-7-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7195
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart