ADV412110985 AChemBlock

4-Amino-1-Cbz-isonipecotic acid

  • MDL number MFCD03425218
  • Cas Number : 115655-41-9
  • Molecular Weight : 278.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-amino-1-cbz-isonipecotic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7204
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart