ADV412110993 AChemBlock

2-Chlorothieno[3,2-d]pyrimidin-4(3H)-one

  • MDL number MFCD19382468
  • Cas Number : 1245811-20-4
  • Molecular Weight : 186.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-chlorothieno-3-2-d-pyrimidin-4-3h-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7212
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart