ADV412110994 AChemBlock

6-Aminomethyl-2-Boc-2-aza-spiro[3.3]heptane

  • MDL number MFCD22419342
  • Cas Number : 1363380-79-3
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-aminomethyl-2-boc-2-aza-spiro-3-3-heptane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7213
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart