ADV412110996 AChemBlock

Ethyl 5-Boc-5-aza-spiro[3.4]octane-2-carboxylate

  • MDL number MFCD17016152
  • Cas Number : 1251022-90-8
  • Molecular Weight : 283.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-5-boc-5-aza-spiro-3-4-octane-2-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7215
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart