ADV412110998 AChemBlock

2-(Boc-amino)-5-aza-spiro[3.4]octane

  • MDL number MFCD22566152
  • Cas Number : 1363381-16-1
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-boc-amino-5-aza-spiro-3-4-octane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7217
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart