ADV412111008 AChemBlock

trans-1,3-Cyclobutanediamine

  • MDL number MFCD22566169
  • Cas Number : 1350753-06-8
  • Molecular Weight : 86.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/trans-1-3-cyclobutanediamine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7229
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart