ADV412111015 AChemBlock

(1R,6S)-3-Boc-3,8-diazabicyclo[4.2.0]octane

  • MDL number MFCD18384927
  • Cas Number : 370882-99-8
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1r-6s-3-boc-3-8-diazabicyclo-4-2-0-octane.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7238
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart