ADV412111017 AChemBlock

3-Azetidinethiol hydrochloride

  • MDL number MFCD22570246
  • Cas Number : 179337-60-1
  • Molecular Weight : 125.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-azetidinethiol-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7240
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart