ADV412111019 AChemBlock

3-(Boc-amino)-4,4-difluoropiperidine

  • MDL number MFCD22566186
  • Cas Number : 1052713-53-7
  • Molecular Weight : 236.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-boc-amino-4-4-difluoropiperidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7242
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart