ADV412111033 AChemBlock

4-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole

  • MDL number MFCD22380249
  • Cas Number : 1022158-35-5
  • Molecular Weight : 281.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-1-tetrahydro-2h-pyran-2-yl-1h-indazole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7257
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart