ADV412111041 AChemBlock

3-Boc-3-azabicyclo[3.2.1]octane-8-carboxylic acid

  • MDL number MFCD17016674
  • Cas Number : 1250997-29-5
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-boc-3-azabicyclo-3-2-1-octane-8-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7265
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart