ADV412111042 AChemBlock

(S)-1-Boc-3-bromomethylpiperidine

  • MDL number MFCD11111933
  • Cas Number : 158406-99-6
  • Molecular Weight : 278.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-1-boc-3-bromomethylpiperidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7266
pictograms GHS07
hstatements H302;H313;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart