ADV412111043 AChemBlock

trans-4-Aminotetrahydropyran-3-ol

  • MDL number MFCD22417050
  • Cas Number : 215940-92-4
  • Molecular Weight : 117.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/trans-4-aminotetrahydropyran-3-ol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7267
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart